Assay Details
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133609.2 | NP_598293.2 | 2 - 3 | 260 | 64 | |
XM_006238630.2 | XP_006238692.1 | 2 - 3 | 231 | 64 | ||
GenBank mRNA | BC072507.1 | - | 2 - 3 | 260 | 7099973 | 64 |
U38253.1 | - | 2 - 3 | 240 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
