Assay Details
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133609.2 | NP_598293.2 | 4 - 5 | 563 | 72 | |
XM_006238630.2 | XP_006238692.1 | 4 - 5 | 534 | 72 | ||
GenBank mRNA | BC072507.1 | - | 4 - 5 | 563 | 72 | |
U38253.1 | - | 4 - 5 | 543 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
