Assay Details
Gene Symbol: Eif2b3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133609.2 | NP_598293.2 | 1 - 2 | 97 | 64 | |
GenBank mRNA | BC072507.1 | - | 1 - 2 | 97 | 7099973 | 64 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC072507.1, NM_133609.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
