Assay Details
Gene Symbol: Zyg11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_233348.8 | XP_233348.6 | 8 - 9 | 1555 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zyg11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_233348.8 | XP_233348.6 | 8 - 9 | 1555 | 141 |
Set Membership: |
![]() ![]() ![]() |