Assay Details
Gene Symbol: Pcsk9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199253.2 | NP_954862.2 | 5 - 6 | 992 | 133 | |
GenBank mRNA | AY847775.1 | - | 5 - 6 | 992 | 133 | |
BC133063.1 | - | 5 - 6 | 923 | 133 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
