Assay Details
Gene Symbol: Urod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019209.1 | NP_062082.1 | 1 - 2 | 58 | 68 | |
GenBank mRNA | BC158690.1 | - | 1 - 2 | 58 | 68 | |
DY314066.1 | - | 1 - 2 | 58 | 68 | ||
FQ234620.1 | - | 1 - 2 | 56 | 68 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC158690.1, NM_019209.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
