Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004236.1 | NP_001004236.1 | 2 - 3 | 187 | 100 | |
XM_006238639.1 | XP_006238701.1 | 4 - 5 | 460 | 100 | ||
GenBank mRNA | BC078938.1 | - | 2 - 3 | 187 | 100 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC078938.1, NM_001004236.1, XM_006238639.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
