Assay Details
Gene Symbol: Tspan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004236.1 | NP_001004236.1 | 6 - 7 | 723 | 89 | |
XM_006238639.1 | XP_006238701.1 | 8 - 9 | 996 | 89 | ||
GenBank mRNA | BC078938.1 | - | 6 - 7 | 723 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
