Assay Details
Gene Symbol: Elavl4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238595.2 | XP_006238657.1 | 6 - 7 | 1043 | 123 | |
XM_008763893.1 | XP_008762115.1 | 6 - 7 | 970 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
