Assay Details
Gene Symbol: Elovl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044275.1 | NP_001037740.1 | 4 - 5 | 468 | 100 | |
GenBank mRNA | BC085795.1 | - | 4 - 5 | 468 | 7109632 | 100 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
