Assay Details
Gene Symbol: Ttc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013214.1 | NP_001013232.1 | 2 - 3 | 242 | 122 | |
XM_006238519.2 | XP_006238581.1 | 2 - 3 | 341 | 122 | ||
GenBank mRNA | BC083794.1 | - | 2 - 3 | 242 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
