Assay Details
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC059133.1 | - | 1 - 2 | 329 | 6918695 | 100 |
FQ212523.1 | - | 1 - 2 | 292 | 100 | ||
FQ212779.1 | - | 1 - 2 | 325 | 100 | ||
FQ213467.1 | - | 1 - 2 | 325 | 100 | ||
FQ213491.1 | - | 1 - 2 | 323 | 100 | ||
FQ213579.1 | - | 1 - 2 | 292 | 100 | ||
FQ214173.1 | - | 1 - 2 | 292 | 100 | ||
FQ214441.1 | - | 1 - 2 | 69 | 100 | ||
FQ214642.1 | - | 307 | 100 | |||
FQ214700.1 | - | 1 - 2 | 265 | 100 | ||
FQ215620.1 | - | 1 - 2 | 117 | 100 | ||
FQ218721.1 | - | 1 - 2 | 292 | 100 | ||
FQ226221.1 | - | 1 - 2 | 325 | 100 | ||
FQ228131.1 | - | 1 - 2 | 325 | 100 | ||
FQ228132.1 | - | 1 - 2 | 325 | 100 | ||
FQ228842.1 | - | 1 - 2 | 321 | 100 | ||
FQ233156.1 | - | 1 - 2 | 325 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC059133.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
