Assay Details
Gene Symbol: Akr1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031000.3 | NP_112262.1 | 5 - 6 | 604 | 105 | |
GenBank mRNA | BC059133.1 | - | 5 - 6 | 883 | 6918695 | 105 |
D10854.1 | - | 5 - 6 | 604 | 105 | ||
FQ212586.1 | - | 5 - 6 | 844 | 105 | ||
FQ212919.1 | - | 4 - 5 | 554 | 105 | ||
FQ213220.1 | - | 4 - 5 | 551 | 105 | ||
FQ213467.1 | - | 5 - 6 | 879 | 105 | ||
FQ213483.1 | - | 5 - 6 | 875 | 105 | ||
FQ213491.1 | - | 5 - 6 | 877 | 105 | ||
FQ213579.1 | - | 5 - 6 | 847 | 106 | ||
FQ213776.1 | - | 5 - 6 | 846 | 105 | ||
FQ214173.1 | - | 5 - 6 | 847 | 105 | ||
FQ214441.1 | - | 5 - 6 | 623 | 105 | ||
FQ214642.1 | - | 860 | 105 | |||
FQ214700.1 | - | 5 - 6 | 820 | 105 | ||
FQ215620.1 | - | 5 - 6 | 671 | 105 | ||
FQ215772.1 | - | 5 - 6 | 846 | 106 | ||
FQ218383.1 | - | 5 - 6 | 847 | 105 | ||
FQ218721.1 | - | 5 - 6 | 846 | 105 | ||
FQ218729.1 | - | 5 - 6 | 846 | 105 | ||
FQ220055.1 | - | 3 - 4 | 447 | 105 | ||
FQ226221.1 | - | 5 - 6 | 879 | 105 | ||
FQ228131.1 | - | 5 - 6 | 880 | 105 | ||
FQ228132.1 | - | 5 - 6 | 879 | 105 | ||
FQ229180.1 | - | 4 - 5 | 518 | 105 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
