Assay Details
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133316.1 | NP_579850.1 | 14 - 15 | 1438 | 79 | |
XM_006238626.2 | XP_006238688.1 | 14 - 15 | 1466 | 79 | ||
XM_006238627.2 | XP_006238689.1 | 13 - 14 | 1363 | 79 | ||
XM_008763987.1 | XP_008762209.1 | 12 - 13 | 1424 | 79 | ||
GenBank mRNA | AF478683.1 | - | 14 - 15 | 1438 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
