Assay Details
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133316.1 | NP_579850.1 | 3 - 4 | 109 | ||
XM_006238626.2 | XP_006238688.1 | 3 - 4 | 344 | 109 | ||
XM_006238627.2 | XP_006238689.1 | 3 - 4 | 355 | 109 | ||
GenBank mRNA | AF478683.1 | - | 3 - 4 | 109 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
