Assay Details
Gene Symbol: Mutyh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133316.1 | NP_579850.1 | 7 - 8 | 539 | 186 | |
XM_006238626.2 | XP_006238688.1 | 7 - 8 | 567 | 186 | ||
XM_008763987.1 | XP_008762209.1 | 5 - 6 | 525 | 287 | ||
GenBank mRNA | AF478683.1 | - | 7 - 8 | 539 | 186 | |
BC128728.1 | - | 7 - 8 | 469 | 186 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
