Assay Details
Gene Symbol: Ppih | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750019.3 | XP_003750067.1 | 1 - 2 | 95 | 73 | |
XM_006238771.2 | XP_006238833.1 | 1 - 2 | 95 | 73 | ||
XM_006238772.2 | XP_006238834.1 | 1 - 2 | 95 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
