Assay Details
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 12 - 13 | 1207 | 96 | |
NM_134472.3 | NP_608302.2 | 11 - 12 | 1092 | 96 | ||
XM_006238632.2 | XP_006238694.1 | 12 - 13 | 1259 | 96 | ||
XM_006238633.1 | XP_006238695.1 | 11 - 12 | 1023 | 96 | ||
XM_006238634.2 | XP_006238696.1 | 11 - 12 | 1555 | 96 | ||
XM_006238635.2 | XP_006238697.1 | 12 - 13 | 1432 | 96 | ||
XM_006238636.2 | XP_006238698.1 | 12 - 13 | 1428 | 96 | ||
GenBank mRNA | CK596636.1 | - | 4 - 5 | 344 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
