Assay Details
Gene Symbol: Kif2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085369.1 | NP_001078838.1 | 10 - 11 | 1044 | 122 | |
NM_134472.3 | NP_608302.2 | 9 - 10 | 929 | 122 | ||
XM_006238632.2 | XP_006238694.1 | 10 - 11 | 1096 | 122 | ||
XM_006238633.1 | XP_006238695.1 | 9 - 10 | 860 | 122 | ||
XM_006238634.2 | XP_006238696.1 | 9 - 10 | 1392 | 122 | ||
XM_006238635.2 | XP_006238697.1 | 10 - 11 | 1269 | 122 | ||
XM_006238636.2 | XP_006238698.1 | 10 - 11 | 1265 | 122 | ||
GenBank mRNA | CK596636.1 | - | 2 - 3 | 181 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
