Assay Details
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005543.1 | NP_001005543.1 | 13 - 14 | 2206 | 141 | |
XM_006238646.2 | XP_006238708.1 | 13 - 14 | 2152 | 141 | ||
XM_006238647.2 | XP_006238709.1 | 11 - 12 | 1188 | 141 | ||
XM_008763993.1 | XP_008762215.1 | 13 - 14 | 2229 | 141 | ||
GenBank mRNA | BC081913.1 | - | 13 - 14 | 2206 | 7112076 | 141 |
FQ212041.1 | - | 12 - 13 | 2146 | 141 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
