Assay Details
Gene Symbol: Nasp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005543.1 | NP_001005543.1 | 4 - 5 | 386 | 73 | |
XM_006238646.2 | XP_006238708.1 | 4 - 5 | 332 | 73 | ||
XM_006238647.2 | XP_006238709.1 | 3 - 4 | 349 | 73 | ||
XM_008763993.1 | XP_008762215.1 | 4 - 5 | 409 | 73 | ||
GenBank mRNA | BC081913.1 | - | 4 - 5 | 386 | 73 | |
FQ212041.1 | - | 3 - 4 | 326 | 73 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
