Assay Details
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238749.2 | XP_006238811.1 | 11 - 12 | 1572 | 75 | |
XM_008764088.1 | XP_008762310.1 | 2725 | 75 | |||
XM_008764089.1 | XP_008762311.1 | 2545 | 75 | |||
XM_345572.8 | XP_345573.5 | 11 - 12 | 1753 | 75 | ||
GenBank mRNA | AB235197.1 | - | 5 - 6 | 607 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
