Assay Details
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238749.2 | XP_006238811.1 | 3 - 4 | 469 | 134 | |
XM_008764089.1 | XP_008762311.1 | 1442 | 134 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
