Assay Details
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238749.2 | XP_006238811.1 | 7 - 8 | 1094 | 85 | |
XM_008764088.1 | XP_008762310.1 | 2247 | 85 | |||
XM_008764089.1 | XP_008762311.1 | 2067 | 85 | |||
XM_008764090.1 | XP_008762312.1 | 7 - 8 | 1281 | 85 | ||
XM_345572.8 | XP_345573.5 | 7 - 8 | 1275 | 85 | ||
GenBank mRNA | AB235197.1 | - | 1 - 2 | 129 | 85 | |
DQ013343.1 | - | 2 - 3 | 274 | 85 | ||
DQ013344.1 | - | 2 - 3 | 274 | 85 | ||
DQ013345.1 | - | 2 - 3 | 274 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
