Assay Details
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238749.2 | XP_006238811.1 | 9 - 10 | 1391 | 99 | |
XM_008764088.1 | XP_008762310.1 | 2544 | 99 | |||
XM_008764089.1 | XP_008762311.1 | 2364 | 99 | |||
XM_345572.8 | XP_345573.5 | 9 - 10 | 1572 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
