Assay Details
Gene Symbol: Mpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008764088.1 | XP_008762310.1 | 1437 | 137 | ||
XM_008764090.1 | XP_008762312.1 | 3 - 4 | 471 | 137 | ||
XM_345572.8 | XP_345573.5 | 3 - 4 | 465 | 137 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
