Assay Details
Gene Symbol: Atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107959.1 | NP_001101429.1 | 4 - 5 | 721 | 111 | |
XM_006238656.2 | XP_006238718.1 | 4 - 5 | 1537 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
