Assay Details
Gene Symbol: Atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107959.1 | NP_001101429.1 | 6 - 7 | 817 | 95 | |
XM_006238656.2 | XP_006238718.1 | 6 - 7 | 1633 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
