Assay Details
Gene Symbol: Atpaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107959.1 | NP_001101429.1 | 8 - 9 | 1018 | 85 | |
XM_006238656.2 | XP_006238718.1 | 8 - 9 | 1834 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P562202 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 791322 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973813 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475013 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 245922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99710 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 68248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 78272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 948158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417551 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 834193 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900582 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 520004 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 220433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 539694 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890103 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 13268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 7658 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 172831 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81852 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P562202 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P562202 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P562202 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P265583 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 16930 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10871119 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871125 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871126 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871122 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871124 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871118 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871121 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871123 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871120 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10871117 | - | - |
Rat Genome 230 2.0 Array | 1373729_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392161_PM_at | - | - |
Rat Genome 230 2.0 Array | 1392161_at | - | - |
Rat Genome 230 2.0 Array | 1373729_at | - | - |
Affymetrix HT Rat Focus Array | 1392161_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392161_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0500008519.rn.1 | TC0500001247.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500008514.rn.1 | TC0500001247.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500008513.rn.1 | TC0500001247.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500008518.rn.1 | TC0500001247.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0500008517.rn.1 | TC0500001247.rn.1 | RTA TC |
Rat Expression Set 230 | 1373729_at | - | - |
Rat Expression Set 230 | 1373729_PM_at | - | - |
Rat Expression Set 230 | 1392161_at | - | - |
Rat Expression Set 230 | 1392161_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10871117 | - | - |
Rat Genome U34 Set | rc_AA997694_at | - | - |
Rat Genome U34 Set | rc_AI030731_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 22033801 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370313 | - | - |
