Assay Details
Gene Symbol: Dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080148.1 | NP_001073617.1 | 2 - 3 | 392 | 65 | |
GenBank mRNA | AY310319.1 | - | 2 - 3 | 392 | 65 | |
AY921220.2 | - | 2 - 3 | 454 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
