Assay Details
Gene Symbol: Dhcr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080148.1 | NP_001073617.1 | 4 - 5 | 617 | 67 | |
GenBank mRNA | AY310319.1 | - | 4 - 5 | 617 | 67 | |
AY921220.2 | - | 4 - 5 | 679 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
