Assay Details
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106680.1 | NP_001100150.1 | 1 - 2 | 184 | 87 | |
XM_006238651.1 | XP_006238713.1 | 1 - 2 | 179 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
