Assay Details
Gene Symbol: Toe1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106680.1 | NP_001100150.1 | 5 - 6 | 619 | 81 | |
XM_006238651.1 | XP_006238713.1 | 5 - 6 | 614 | 81 | ||
XM_006238652.2 | XP_006238714.1 | 4 - 5 | 944 | 81 | ||
XM_006238653.2 | XP_006238715.1 | 4 - 5 | 866 | 81 | ||
XM_006238654.2 | XP_006238716.1 | 4 - 5 | 967 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
