Assay Details
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107965.1 | NP_001101435.1 | 4 - 5 | 922 | 67 | |
XM_006238667.2 | XP_006238729.1 | 4 - 5 | 992 | 67 | ||
GenBank mRNA | FQ214130.1 | - | 2 - 3 | 413 | 67 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
