Assay Details
Gene Symbol: B4galt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107965.1 | NP_001101435.1 | 5 - 6 | 1048 | 56 | |
XM_006238667.2 | XP_006238729.1 | 5 - 6 | 1118 | 56 | ||
GenBank mRNA | FQ214130.1 | - | 3 - 4 | 539 | 56 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
