Assay Details
Gene Symbol: Ccdc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006238709.2 | XP_006238771.1 | 6 - 7 | 956 | 103 | |
XM_006238710.2 | XP_006238772.1 | 6 - 7 | 944 | 103 | ||
XM_008764052.1 | XP_008762274.1 | 6 - 7 | 986 | 103 | ||
XM_008764053.1 | XP_008762275.1 | 6 - 7 | 974 | 103 | ||
XM_008764054.1 | XP_008762276.1 | 6 - 7 | 986 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
