Assay Details
Gene Symbol: Prpf38a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106672.1 | NP_001100142.1 | 5 - 6 | 808 | 69 | |
GenBank mRNA | FQ221356.1 | - | 3 - 4 | 214 | 69 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
