Assay Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013109.1 | NP_001013127.1 | 6 - 7 | 578 | 78 | |
XM_006243306.2 | XP_006243368.1 | 10 - 11 | 1095 | 78 | ||
GenBank mRNA | BC091410.1 | - | 6 - 7 | 578 | 78 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
