Assay Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243306.2 | XP_006243368.1 | 3 - 4 | 463 | 71 | |
XM_008766241.1 | XP_008764463.1 | 3 - 4 | 473 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
