Assay Details
Gene Symbol: Dapk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013109.1 | NP_001013127.1 | 5 - 6 | 493 | 99 | |
XM_006243306.2 | XP_006243368.1 | 9 - 10 | 1010 | 99 | ||
GenBank mRNA | BC091410.1 | - | 5 - 6 | 493 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
