Assay Details
Gene Symbol: Mfrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108137.2 | NP_001101607.1 | 12 - 13 | 1704 | 75 | |
XM_006242933.2 | XP_006242995.1 | 12 - 13 | 2131 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
