Assay Details
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053297.2 | NP_445749.1 | 6 - 7 | 912 | 66 | |
XM_006243187.2 | XP_006243249.1 | 6 - 7 | 1142 | 66 | ||
XM_006243188.2 | XP_006243250.1 | 6 - 7 | 1142 | 66 | ||
XM_006243190.2 | XP_006243252.1 | 7 - 8 | 1214 | 66 | ||
GenBank mRNA | AY724474.1 | - | 6 - 7 | 909 | 66 | |
FQ223721.1 | - | 6 - 7 | 920 | 66 | ||
FQ224316.1 | - | 3 - 4 | 570 | 66 | ||
X15800.1 | - | 6 - 7 | 914 | 66 |
Gene Symbol: LOC100362738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772937.1 | XP_008771159.1 | 1 | 876 | 66 |
Gene Symbol: LOC100364062 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751952.3 | XP_003752000.1 | 1 | 836 | 66 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
