Assay Details
Gene Symbol: Pkm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053297.2 | NP_445749.1 | 7 - 8 | 1068 | 113 | |
XM_006243187.2 | XP_006243249.1 | 7 - 8 | 1298 | 113 | ||
XM_006243188.2 | XP_006243250.1 | 7 - 8 | 1298 | 113 | ||
XM_006243190.2 | XP_006243252.1 | 8 - 9 | 1370 | 113 | ||
GenBank mRNA | AY724474.1 | - | 7 - 8 | 1065 | 113 | |
FQ223721.1 | - | 7 - 8 | 1076 | 113 | ||
FQ224316.1 | - | 4 - 5 | 726 | 113 | ||
X15800.1 | - | 7 - 8 | 1070 | 113 |
Gene Symbol: LOC100362738 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008772937.1 | XP_008771159.1 | 1 | 1032 | 113 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
