Assay Details
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007803.1 | NP_001007804.1 | 11 - 12 | 1607 | 88 | |
XM_006243250.1 | XP_006243312.1 | 10 - 11 | 1810 | 88 | ||
GenBank mRNA | BC085867.1 | - | 11 - 12 | 1733 | 88 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
