Assay Details
Gene Symbol: Clpx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007803.1 | NP_001007804.1 | 1 - 2 | 77 | 95 | |
XM_006243250.1 | XP_006243312.1 | 1 - 2 | 322 | 95 | ||
GenBank mRNA | BC085867.1 | - | 1 - 2 | 203 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
