Assay Details
Gene Symbol: Chrna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_052805.2 | NP_434692.2 | 3 - 4 | 240 | 135 | |
GenBank mRNA | AY574254.1 | - | 3 - 4 | 240 | 135 | |
L31621.1 | - | 3 - 4 | 316 | 135 | ||
X03440.1 | - | 3 - 4 | 390 | 135 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
