Assay Details
Gene Symbol: Csk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030039.1 | NP_001025210.1 | 6 - 7 | 669 | 119 | |
XM_006243163.1 | XP_006243225.1 | 7 - 8 | 1315 | 119 | ||
XM_006243164.2 | XP_006243226.1 | 7 - 8 | 998 | 119 | ||
GenBank mRNA | BC098863.1 | - | 6 - 7 | 669 | 7380519 | 119 |
X58631.1 | - | 7 - 8 | 704 | 119 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
