Assay Details
Gene Symbol: Ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766429.1 | XP_008764651.1 | 4 - 5 | 786 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
