Assay Details
Gene Symbol: Ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766429.1 | XP_008764651.1 | 6 - 7 | 1006 | 80 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
