Assay Details
Gene Symbol: Ube2q2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766429.1 | XP_008764651.1 | 8 - 9 | 1153 | 142 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
